(S)-2-(Dimethylcarbamothioyl)-N-(2-methyl-4-(1-methyl-1,4,5,10-tetrahydrobenzo[b]pyrazolo[3,4-e][1,4]diazepine-5-carbonyl)benzyl)pyrrolidine-1-carboxamide 2,2,2-trifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: LIT-001

A1207046-10MG A1207046-1MG A1207046-25MG A1207046-50MG A1207046-5MG
76677-672EA 91.24 USD
76677-672 76677-674 76677-676 76677-678 76677-680
(S)-2-(Dimethylcarbamothioyl)-N-(2-methyl-4-(1-methyl-1,4,5,10-tetrahydrobenzo[b]pyrazolo[3,4-e][1,4]diazepine-5-carbonyl)benzyl)pyrrolidine-1-carboxamide 2,2,2-trifluoroacetate 98%
(S)-2-(Dimethylcarbamothioyl)-N-(2-methyl-4-(1-methyl-1,4,5,10-tetrahydrobenzo[b]pyrazolo[3,4-e][1,4]diazepine-5-carbonyl)benzyl)pyrrolidine-1-carboxamide 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₃₀H₃₄F₃N₇O₄S
MW: 645.7 g/mol
MDL Number: MFCD32644853
CAS Number: 2245072-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR