2,6-Dibromo-3,4,5-trimethylaniline 97%

Supplier: Ambeed
Warning

A1207022-100MG A1207022-1G A1207022-250MG A1207022-5G
76677-666EA 7 USD
76677-666 76677-668 76677-670 77596-246
2,6-Dibromo-3,4,5-trimethylaniline 97%
2,6-Dibromo-3,4,5-trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₁Br₂N
MDL Number: MFCD32856208
CAS Number: 68818-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR