tert-Butyl 2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoyl)hydrazinecarboxylate 98%

Supplier: Ambeed
Danger

A1206964-1G A1206964-250MG
76677-660EA 113.52 USD
76677-660 76677-662
tert-Butyl 2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoyl)hydrazinecarboxylate 98%
tert-Butyl 2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoyl)hydrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₅
MW: 283.28 g/mol
MDL Number: MFCD09840851
CAS Number: 1076198-38-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR