5-Bromo-2-hydrazinylbenzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A1206782-100MG A1206782-250MG
76677-654EA 15 USD
76677-654 76677-658
5-Bromo-2-hydrazinylbenzoic acid hydrochloride 97%
5-Bromo-2-hydrazinylbenzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈BrClN₂O₂
MDL Number: MFCD10000943
CAS Number: 1260776-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR