1-(3-Iodobenzyl)guanidine hemisulfate 97%

Supplier: Ambeed
Warning

A1206320-1G A1206320-250MG A1206320-5G
76677-650EA 492 USD
76677-650 76677-652 77620-814
1-(3-Iodobenzyl)guanidine hemisulfate 97%
1-(3-Iodobenzyl)guanidine hemisulfate

Precaución: for Research Use Only

Formula: C₁₆H₂₂I₂N₆O₄S
MDL Number: MFCD00151189
CAS Number: 87862-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR