Ethyl 2-(pyridin-4-yloxy)acetate 98%
Supplier: Ambeed
|
Warning
|
A1206222-1G
A1206222-5G
76677-646EA
89.12
USD
76677-646
76677-648
Ethyl 2-(pyridin-4-yloxy)acetate 98%
Ethyl 2-(pyridin-4-yloxy)acetate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₃ MW: 181.1885 g/mol |
CAS Number:
58530-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...