2-Chloro-3-(methoxymethoxy)pyridine 95%

Supplier: Ambeed
Danger

A1206138-1G A1206138-250MG A1206138-5G
76677-636EA 78.51 USD
76677-636 76677-638 76677-640
2-Chloro-3-(methoxymethoxy)pyridine 95%
2-Chloro-3-(methoxymethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.5969 g/mol
MDL Number: MFCD21195353
CAS Number: 862667-72-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR