[1,1'-binaphthalene]-2,2'-diylbis(diphenylphosphine oxide) 98%

Supplier: Ambeed
Warning

A1206081-1G A1206081-5G
76677-632EA 16.98 USD
76677-632 76677-634
[1,1'-binaphthalene]-2,2'-diylbis(diphenylphosphine oxide) 98%
[1,1'-binaphthalene]-2,2'-diylbis(diphenylphosphine oxide)

Precaución: for Research Use Only

Formula: C₄₄H₃₂O₂P₂
MW: 654.6712 g/mol
CAS Number: 86632-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR