Diethyl benzhydrylphosphonate 98%

Supplier: Ambeed
Warning

A1206009-5G A1206009-25G
76677-624EA 9 USD
76677-624 77596-240
Diethyl benzhydrylphosphonate 98%
Diethyl benzhydrylphosphonate

Precaución: for Research Use Only

Formula: C₁₇H₂₁O₃P
MDL Number: MFCD00143661
CAS Number: 27329-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR