2-(3,5-Dimethylphenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1205954-1G A1205954-250MG A1205954-5G
76677-618EA 26.53 USD
76677-618 76677-620 76677-622
2-(3,5-Dimethylphenyl)-1,3,2-dioxaborinane 98%
2-(3,5-Dimethylphenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₂
MW: 190.0466 g/mol
CAS Number: 436853-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR