2-Chloro-4-(dibenzo[b,d]furan-1-yl)-6-phenyl-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A1205859-1G A1205859-250MG A1205859-5G
76677-612EA 28.65 USD
76677-612 76677-614 76677-616
2-Chloro-4-(dibenzo[b,d]furan-1-yl)-6-phenyl-1,3,5-triazine 98%
2-Chloro-4-(dibenzo[b,d]furan-1-yl)-6-phenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₂ClN₃O
MW: 357.7925 g/mol
MDL Number: MFCD32709292
CAS Number: 1883265-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR