2,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A1205858-100MG A1205858-250MG A1205858-50MG
76677-606EA 59 USD
76677-606 76677-608 76677-610
2,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₃₅H₂₇N₃O₂
MDL Number: MFCD31706435
CAS Number: 410092-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR