Thianthrene 5-oxide 97%

Supplier: Ambeed
Warning

A1205571-100MG A1205571-250MG A1205571-1G A1205571-5G
76677-596EA 39 USD
76677-596 76677-598 77592-338 77592-340
Thianthrene 5-oxide 97%
Thianthrene 5-oxide

Precaución: for Research Use Only

Formula: C₁₂H₈OS₂
MDL Number: MFCD01464038
CAS Number: 2362-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR