6-(2-(Methylthio)pyrimidin-5-yl)hex-5-ynoic acid 98%

Supplier: Ambeed
Warning

A1205379-100MG A1205379-1G A1205379-250MG A1205379-5G
76677-586EA 97.61 USD
76677-586 76677-588 76677-590 76677-592
6-(2-(Methylthio)pyrimidin-5-yl)hex-5-ynoic acid 98%
6-(2-(Methylthio)pyrimidin-5-yl)hex-5-ynoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂S
MW: 236.2902 g/mol
CAS Number: 2356229-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR