2-Bromo-1-(6-(difluoromethoxy)pyridin-3-yl)ethanone 98%

Supplier: Ambeed
Danger

A1205212-100MG A1205212-250MG
76677-582EA 92.3 USD
76677-582 76677-584
2-Bromo-1-(6-(difluoromethoxy)pyridin-3-yl)ethanone 98%
2-Bromo-1-(6-(difluoromethoxy)pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrF₂NO₂
MW: 266.0396 g/mol
MDL Number: MFCD28659190
CAS Number: 1523194-44-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR