tert-Butyl (1-(6-chloropyridin-3-yl)cyclobutyl)carbamate 98%

Supplier: Ambeed
Warning

A1205207-100MG A1205207-250MG
76677-578EA 43 USD
76677-578 76677-580
tert-Butyl (1-(6-chloropyridin-3-yl)cyclobutyl)carbamate 98%
tert-Butyl (1-(6-chloropyridin-3-yl)cyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₉ClN₂O₂
MDL Number: MFCD28400598
CAS Number: 1887059-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR