N,5-Dimethylpyridin-3-amine 98%
Supplier: Ambeed
|
Warning
|
A1205040-250MG
A1205040-1G
76677-576EA
112.46
USD
76677-576
77620-800
N,5-Dimethylpyridin-3-amine 98%
N,5-Dimethylpyridin-3-amine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ MW: 122.1677 g/mol |
MDL Number:
MFCD28122622 CAS Number: 1610612-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...