2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)benzene-1-sulfonyl chloride 97%

Supplier: Ambeed
Danger

A1204927-25G A1204927-5G
76677-570EA 360.71 USD
76677-570 76677-572
2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)benzene-1-sulfonyl chloride 97%
2-(2,2-Difluoroethoxy)-6-(trifluoromethyl)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₆ClF₅O₃S
MW: 324.6522 g/mol
MDL Number: MFCD26142688
CAS Number: 865352-01-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR