3-Chloro-2-(3-(trifluoromethyl)phenoxy)pyridine 97%

Supplier: Ambeed
Warning

A1204923-1G A1204923-250MG A1204923-5G
76677-564EA 29 USD
76677-564 76677-566 76677-568
3-Chloro-2-(3-(trifluoromethyl)phenoxy)pyridine 97%
3-Chloro-2-(3-(trifluoromethyl)phenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₇ClF₃NO
CAS Number: 197565-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR