6,7-Dimethoxy-N-(3-phenoxyphenyl)quinolin-4-amine hydrochloride 98%

Supplier: Ambeed
Warning

A1204840-10MG A1204840-1MG A1204840-50MG A1204840-5MG
76677-548EA 33.95 USD
76677-548 76677-550 76677-552 76677-554
6,7-Dimethoxy-N-(3-phenoxyphenyl)quinolin-4-amine hydrochloride 98%
6,7-Dimethoxy-N-(3-phenoxyphenyl)quinolin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₁ClN₂O₃
MW: 408.88 g/mol
CAS Number: 179246-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR