6-Bromo-2,3-dihydro-1H-inden-1-amine 98%

Supplier: Ambeed
Warning

A1204806-250MG A1204806-100MG A1204806-5G
76677-546EA 54.11 USD
76677-546 77620-796 77620-798
6-Bromo-2,3-dihydro-1H-inden-1-amine 98%
6-Bromo-2,3-dihydro-1H-inden-1-amine

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
MW: 212.0864 g/mol
MDL Number: MFCD07373953
CAS Number: 907973-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR