Methyl 3-hydroxy-5-methoxy-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A1204359-1G A1204359-250MG A1204359-5G
76677-524EA 114.58 USD
76677-524 76677-526 76677-528
Methyl 3-hydroxy-5-methoxy-1H-indole-2-carboxylate 98%
Methyl 3-hydroxy-5-methoxy-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₄
MW: 221.2093 g/mol
MDL Number: MFCD27965020
CAS Number: 70258-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR