2-Bromo-5-(piperidin-4-yl)pyridine 97%

Supplier: Ambeed
Danger

A1197747-100MG A1197747-1G A1197747-250MG A1197747-5G
76677-512EA 96.55 USD
76677-512 76677-514 76677-516 76677-518
2-Bromo-5-(piperidin-4-yl)pyridine 97%
2-Bromo-5-(piperidin-4-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂
MW: 241.1276 g/mol
MDL Number: MFCD12033501
CAS Number: 1159814-58-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR