2-(2-((6-Chlorohexyl)oxy)ethoxy)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(6-chlorohexyloxy)ethoxy)ethanamine HCl

A1197707-100MG A1197707-250MG A1197707-1G A1197707-5G
76677-502EA 95.49 USD
76677-502 76677-504 77549-404 77571-192
2-(2-((6-Chlorohexyl)oxy)ethoxy)ethanamine hydrochloride 97%
2-(2-((6-Chlorohexyl)oxy)ethoxy)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₃Cl₂NO₂
MW: 260.2011 g/mol
MDL Number: MFCD32706550
CAS Number: 1035373-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR