tert-Butyl ((3S,4S)-1-chloro-4-methyl-2-oxohexan-3-yl)carbamate 98%

Supplier: Ambeed
Warning

A1196551-100MG A1196551-250MG
76677-476EA 39.26 USD
76677-476 76677-478
tert-Butyl ((3S,4S)-1-chloro-4-methyl-2-oxohexan-3-yl)carbamate 98%
tert-Butyl ((3S,4S)-1-chloro-4-methyl-2-oxohexan-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₂ClNO₃
MW: 263.7610 g/mol
CAS Number: 161805-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR