2,2-Dimethyl-1-(2-methylquinolin-6-yl)propan-1-one 97%

Supplier: Ambeed
Warning

A1196071-1G A1196071-250MG
76677-472EA 321.46 USD
76677-472 76677-474
2,2-Dimethyl-1-(2-methylquinolin-6-yl)propan-1-one 97%
2,2-Dimethyl-1-(2-methylquinolin-6-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO
MW: 227.3016 g/mol
MDL Number: MFCD23537048
CAS Number: 1534944-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR