6,6',6'',6'''-(Pyrene-1,3,6,8-tetrayl)tetrakis(2-naphthoic acid) 97%

Supplier: Ambeed
Warning

A1195942-100MG A1195942-1G A1195942-250MG
76677-456EA 54.11 USD
76677-456 76677-458 76677-460
6,6',6'',6'''-(Pyrene-1,3,6,8-tetrayl)tetrakis(2-naphthoic acid) 97%
6,6',6'',6'''-(Pyrene-1,3,6,8-tetrayl)tetrakis(2-naphthoic acid)

Precaución: for Research Use Only

Formula: C₆₀H₃₄O₈
MW: 882.907 g/mol
MDL Number: MFCD34563557
CAS Number: 2010983-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR