4,4'-((2,5-Dimethoxy-1,4-phenylene)bis(ethyne-2,1-diyl))dibenzaldehyde 98%

Supplier: Ambeed
Warning

A1195934-100MG A1195934-1G A1195934-250MG
76677-444EA 56.23 USD
76677-444 76677-446 76677-448
4,4'-((2,5-Dimethoxy-1,4-phenylene)bis(ethyne-2,1-diyl))dibenzaldehyde 98%
4,4'-((2,5-Dimethoxy-1,4-phenylene)bis(ethyne-2,1-diyl))dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₆H₁₈O₄
MW: 394.4187 g/mol
MDL Number: MFCD32667011
CAS Number: 1190963-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR