4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carbonitrile)) 98%
Supplier: Ambeed
|
Warning
|
A1195908-100MG
A1195908-1G
A1195908-250MG
76677-438EA
56.23
USD
76677-438
76677-440
76677-442
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carbonitrile)) 98%
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carbonitrile))
Precaución: for Research Use Only
Formula:
C₃₉H₂₄N₄ MW: 548.6347 g/mol |
MDL Number:
MFCD32696917 CAS Number: 1800552-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...