1,4-Bis((2-methyl-1H-imidazol-1-yl)methyl)benzene 98%
Supplier: Ambeed
|
Warning
|
A1195894-1G
A1195894-250MG
A1195894-5G
76677-432EA
127.31
USD
76677-432
76677-434
76677-436
1,4-Bis((2-methyl-1H-imidazol-1-yl)methyl)benzene 98%
1,4-Bis((2-methyl-1H-imidazol-1-yl)methyl)benzene
Precaución: for Research Use Only
Formula:
C₁₆H₁₈N₄ MW: 266.3400 g/mol |
CAS Number:
82410-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...