5-(Pyridin-4-ylmethoxy)isophthalic acid 98%

Supplier: Ambeed
Warning

A1195893-100MG A1195893-1G A1195893-250MG
76677-426EA 18.04 USD
76677-426 76677-428 76677-430
5-(Pyridin-4-ylmethoxy)isophthalic acid 98%
5-(Pyridin-4-ylmethoxy)isophthalic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₅
MW: 273.2408 g/mol
CAS Number: 1240327-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR