4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzaldehyde 98%

Supplier: Ambeed
Warning

A1195877-100MG A1195877-1G A1195877-250MG
76677-410EA 109.28 USD
76677-410 76677-412 76677-414
4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzaldehyde 98%
4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzaldehyde

Precaución: for Research Use Only

Formula: C₆₂H₃₈O₆
MW: 878.9615 g/mol
CAS Number: 1835723-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR