1,1,2,2-Tetrakis(4-(pyridin-3-yl)phenyl)ethene 98%

Supplier: Ambeed
Warning

A1195831-1G A1195831-250MG
76677-402EA 239.77 USD
76677-402 76677-404
1,1,2,2-Tetrakis(4-(pyridin-3-yl)phenyl)ethene 98%
1,1,2,2-Tetrakis(4-(pyridin-3-yl)phenyl)ethene

Precaución: for Research Use Only

Formula: C₄₆H₃₂N₄
MW: 640.7731 g/mol
CAS Number: 1287777-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR