Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methane 98%
Supplier: Ambeed
|
Warning
|
A1195824-100MG
A1195824-1G
A1195824-250MG
A1195824-5G
76677-394EA
54.11
USD
76677-394
76677-396
76677-398
76677-400
Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methane 98%
Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methane
Precaución: for Research Use Only
Formula:
C₄₉H₆₄B₄O₈ MW: 824.2717 g/mol |
CAS Number:
875772-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...