4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A1195820-100MG
A1195820-250MG
A1195820-1G
76677-384EA
28.65
USD
76677-384
76677-386
77596-236
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzoic acid 98%
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₁₂N₂O₄S MW: 376.3853 g/mol |
CAS Number:
1581774-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...