3,3''-Difluoro-5'-(3-fluoro-4-formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1195811-100MG A1195811-1G A1195811-250MG
76677-372EA 21 USD
76677-372 76677-374 76677-376
3,3''-Difluoro-5'-(3-fluoro-4-formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde 98%
3,3''-Difluoro-5'-(3-fluoro-4-formylphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₂₇H₁₅F₃O₃
MW: 444.4014 g/mol
MDL Number: MFCD32667000
CAS Number: 2088007-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR