4-(10,15,20-Triphenylporphyrin-5-yl)phenol 98%

Supplier: Ambeed
Warning

A1195804-100MG A1195804-1G A1195804-250MG A1195804-50MG
76677-364EA 251.44 USD
76677-364 76677-366 76677-368 76677-370
4-(10,15,20-Triphenylporphyrin-5-yl)phenol 98%
4-(10,15,20-Triphenylporphyrin-5-yl)phenol

Precaución: for Research Use Only

Formula: C₄₄H₃₀N₄O
MW: 630.7352 g/mol
MDL Number: MFCD09038998
CAS Number: 87345-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR