4,4''-Di(dihydroxyboryl)-5'-(4-dihydroxy boryl phenyl)-1,1':3',1''-terphenyl 98%

Supplier: Ambeed
Warning

A1195791-100MG A1195791-1G A1195791-250MG
76677-342EA 39.26 USD
76677-342 76677-344 76677-346
4,4''-Di(dihydroxyboryl)-5'-(4-dihydroxy boryl phenyl)-1,1':3',1''-terphenyl 98%
4,4''-Di(dihydroxyboryl)-5'-(4-dihydroxy boryl phenyl)-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₄H₂₁B₃O₆
MW: 437.8530 g/mol
MDL Number: MFCD32667022
CAS Number: 900795-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR