2,5-Bis(octyloxy)terephthalaldehyde 98%

Supplier: Ambeed
Warning

A1195788-1G A1195788-5G
76677-338EA 266.29 USD
76677-338 76677-340
2,5-Bis(octyloxy)terephthalaldehyde 98%
2,5-Bis(octyloxy)terephthalaldehyde

Precaución: for Research Use Only

Formula: C₂₄H₃₈O₄
MW: 390.5561 g/mol
MDL Number: MFCD02063373
CAS Number: 123440-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR