2,5-Bis(hexyloxy)terephthalaldehyde 98%

Supplier: Ambeed
Warning

A1195787-100MG A1195787-250MG A1195787-5G
76677-332EA 38.2 USD
76677-332 76677-334 76677-336
2,5-Bis(hexyloxy)terephthalaldehyde 98%
2,5-Bis(hexyloxy)terephthalaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₃₀O₄
MW: 334.4498 g/mol
MDL Number: MFCD03427236
CAS Number: 151903-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR