4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-3-(trifluoromethyl)phenylboronic acid pinacol ester

A1195390-1G A1195390-250MG A1195390-5G
76677-324EA 128.37 USD
76677-324 76677-326 76677-328
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₆BF₃O₄
MW: 316.08 g/mol
MDL Number: MFCD28356850
CAS Number: 2121513-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR