20-Hydroxyicosanoic acid 98%

Supplier: Ambeed
Warning

A1195096-100MG A1195096-1G A1195096-250MG A1195096-25G A1195096-5G
76677-306EA 15 USD
76677-306 76677-308 76677-310 76677-312 76677-314
20-Hydroxyicosanoic acid 98%
20-Hydroxyicosanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₄₀O₃
MDL Number: MFCD02259050
CAS Number: 62643-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR