Fucoidan 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3S,4S,5S,6R)-4,5-Dihydroxy-2,6-dimethyltetrahydro-2H-pyran-3-yl hydrogen sulphate

A1195062-100G A1195062-25G A1195062-5G
76677-300EA 205 USD
76677-300 76677-302 76677-304
Fucoidan 97%
Fucoidan

Precaución: for Research Use Only

Formula: C₇H₁₄O₇S
MDL Number: MFCD00131109
CAS Number: 9072-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR