Methyl 3-chloro-5-(N-(4,6-difluoro-[1,1'-biphenyl]-3-yl)sulfamoyl)-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: NDI-091143

A1194991-10MG A1194991-50MG A1194991-5MG A1194991-1MG A1194991-25MG
76677-286EA 50.93 USD
76677-286 76677-288 76677-290 77596-224 77620-742
Methyl 3-chloro-5-(N-(4,6-difluoro-[1,1'-biphenyl]-3-yl)sulfamoyl)-4-hydroxybenzoate 98%
Methyl 3-chloro-5-(N-(4,6-difluoro-[1,1'-biphenyl]-3-yl)sulfamoyl)-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClF₂NO₅S
MW: 453.84 g/mol
CAS Number: 2375840-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR