2,2'-(Diazene-1,2-diyl)dibenzoic acid 98%

Supplier: Ambeed
Warning

A1194679-100MG A1194679-1G A1194679-250MG
76677-270EA 93.36 USD
76677-270 76677-272 76677-274
2,2'-(Diazene-1,2-diyl)dibenzoic acid 98%
2,2'-(Diazene-1,2-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₄
MW: 270.2402 g/mol
CAS Number: 635-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR