Allylpalladium(II) chloride dimer 98,5% Pd 56.7-59.6%

Supplier: Ambeed
Warning

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

A1194530-100MG A1194530-250MG
76677-260EA 36.08 USD
76677-260 76677-262
Allylpalladium(II) chloride dimer 98,5% Pd 56.7-59.6%
Allylpalladium(II) chloride dimer

Precaución: for Research Use Only

Formula: C₆H₁₀Cl₂Pd₂
MW: 365.89 g/mol
MDL Number: MFCD00044874
CAS Number: 12012-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR