fac-Tris(2-phenylpyridine)iridium 99%, purified by sublimation

Supplier: Ambeed
Warning

Synonyms: fac-Ir(ppy)3

A1194524-100MG A1194524-1G A1194524-250MG
76677-254EA 46 USD
76677-254 76677-256 76677-258
fac-Tris(2-phenylpyridine)iridium 99%, purified by sublimation
fac-Tris(2-phenylpyridine)iridium

Precaución: for Research Use Only

Formula: C₃₃H₂₄IrN₃
MW: 654.78 g/mol
MDL Number: MFCD12022527
CAS Number: 94928-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR