Palladium(II) acetate 99.9%
Supplier: Ambeed
|
Danger
|
Synonyms:
Palladium diacetate, Palladium (II) acetate
A1194522-10G
A1194522-1G
A1194522-250MG
A1194522-25G
A1194522-5G
76677-244EA
725
USD
76677-244
76677-246
76677-248
76677-250
76677-252
Palladium(II) acetate 99.9%
Palladium(II) acetate
Precaución: for Research Use Only
Formula:
C₄H₆O₄Pd MW: 224.51 g/mol Melting Pt: 205 |
MDL Number:
MFCD00012453 CAS Number: 3375-31-3 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...