BrettPhos palladacycle 3rd generation 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
[(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulphonate, BrettPhos Pd G3
A1194516-100MG
A1194516-250MG
76677-240EA
15.92
USD
76677-240
76677-242
BrettPhos palladacycle 3rd generation 98%
BrettPhos palladacycle 3rd generation
Precaución: for Research Use Only
Formula:
C₄₈H₆₆NO₅PPdS MW: 906.50 g/mol |
MDL Number:
MFCD22572666 CAS Number: 1470372-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...