N-(tert-Butyl)-2-((4,5-diphenyloxazol-2-yl)thio)propanamide 97%

Supplier: Ambeed
Warning

A1194393-100MG A1194393-1G A1194393-250MG
76677-230EA 46.68 USD
76677-230 76677-232 76677-234
N-(tert-Butyl)-2-((4,5-diphenyloxazol-2-yl)thio)propanamide 97%
N-(tert-Butyl)-2-((4,5-diphenyloxazol-2-yl)thio)propanamide

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₂O₂S
MW: 380.5032 g/mol
MDL Number: MFCD15659365
CAS Number: 1385506-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR